missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-2-[(tert-Butyldimethylsilyl)oxy]-2-(3-chlorophenyl)acetate, TRC

Product Code. 30477691
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477691 500 mg 500mg
30479834 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477691 Supplier Toronto Research Chemicals Supplier No. TRCM305188500MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl (R)-2-[(tert-Butyldimethylsilyl)oxy]-2-(3-chlorophenyl)acetate
CAS 153294-01-0
Molecular Formula C15H23ClO3Si
Quantity 500 mg
Synonym (alphaR)-3-Chloro-alpha-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-benzeneacetic Acid Methyl Ester, (R)- 3-Chloro-alpha-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-benzeneacetic Acid Methyl Ester
InChI Formula InChI=1S/C15H23ClO3Si/c1-15(2,3)20(5,6)19-13(14(17)18-4)11-8-7-9-12(16)10-11/h7-10,13H,1-6H3/t13-/m1/s1
SMILES COC(=O)[C@H](O[Si](C)(C)C(C)(C)C)c1cccc(Cl)c1
IUPAC Name methyl (2R)-2-[tert-butyl(dimethyl)silyl]oxy-2-(3-chlorophenyl)acetate
Molecular Weight (g/mol) 314.88
Formula Weight 314.1105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.