missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(-)-β-hydroxyisobutyrate, 98%, Thermo Scientific Chemicals

Product Code. 10431050 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10431050 1 g Glass bottle 1g
10351630 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10431050 Supplier Thermo Scientific Acros Supplier No. 436180010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72657-23-9
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00063450
InChI Key ATCCIZURPPEVIZ-SCSAIBSYSA-N
Synonym r-methyl 3-hydroxy-2-methylpropanoate, r---3-hydroxy-2-methylpropionic acid methyl ester, methyl r---3-hydroxy-2-methylpropionate, --methyl d-beta-hydroxyisobutyrate, methyl r---3-hydroxyisobutyrate, roche ester, methyl r-3-hydroxy-2-methylpropionate, propanoic acid, 3-hydroxy-2-methyl-, methyl ester, 2r, methyl 2r-3-hydroxy-2-methylpropanoate, r---3-hydroxyisobutyric acid methyl ester
PubChem CID 5324733
IUPAC Name methyl (2R)-3-hydroxy-2-methylpropanoate
SMILES CC(CO)C(=O)OC

Specifications

Color Colorless
Density 1.0700g/mL
Boiling Point 60.0°C (4.0 Torr)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2CH(CH3)COOCH3
Refractive Index 1.4243
Quantity 1 g
Specific Gravity 1.07
Specific Rotation − 23.30
Solubility Information Solubility in water: insoluble. Other solubilities: in ethanol: soluble
Formula Weight 118.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl (R)-(-)-β-hydroxyisobutyrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.