missing translation for 'onlineSavingsMsg'
Learn More

Methyl red, For ACS analysis, MP Biomedicals™

Product Code. 11482872 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482872 25 g 25g
11492872 100 g 100g
11402882 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482872 Supplier MP Biomedicals Supplier No. 0215254525
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym 2-(4-Dimethylaminophenylazo)benzoic acid, 4-Dimethylaminoazobenzene-2'-carboxylic acid
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Formula Weight 269.3
Color Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Methyl red, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.