missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red Indicator, Reag. Ph. Eur., Solstice

Product Code. 15623830
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15623830 100 g Glass Bottle 100g
15633830 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15623830 Supplier Honeywell-Fluka Supplier No. 32654100G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reag. Ph. Eur., acid-base indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number MFCD00002425
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym Acid Red 2
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 179°C to 182°C
Linear Formula (CH3)2NC6H4N=NC6H4CO2H
UN Number NONH for all modes of transport
Beilstein 750102
Solubility Information Slightly soluble in water
Formula Weight 269.3g/mol
Packaging Glass Bottle
Suitability Complies for sensitivity
Visual Transition Interval 4.4 to 6.0
Density 0.989g/cm3
Color Violet
Quantity 100 g
Physical Form Powder
Chemical Name or Material Methyl Red, Reag. Ph. Eur.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.