missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red, Pure, Water Soluble, Indicator, Fisher Chemical™

Product Code. 10375710 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10375710 5 g 5g
10793991 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10375710 Supplier Fisher Chemical Supplier No. M/5170/44

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number 2425
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 178°C
Formula Weight 269.3g/mol
Packaging Glass Jar
Color Red
Quantity 5 g
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.