missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Methyl Red is an azo dye that is a pH indicator that turns color in the presence of acid solutions (red in pH <4.4, yellow in pH >6.2, orange in between). It is particularly used in the MR Test for identifying enteric bacteria that produce stable acids during glucose metabolism (fermentation).
- Prepared by mixing 50g methyl red in a mixture of 1.86mL of 0.1M sodium hydroxide and 50mL ethanol, and diluted to 100mL with water
- Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
- Provided with a certificate of analysis and a label that shows the lot and expiration date
- Ready to use reagent eliminates the time and labor of preparing precise volumetric solutions
Specifications
Specifications
| Chemical Name or Material | 2-(4-Dimethylamino-phenylazo)benzoic acid |
| CAS | 493-52-7, 1310-73-2, 64-17-5, 7732-18-5 |
| Concentration | 50mg Methyl Red per 100mL |
| Packaging | Temper evident |
| Molecular Formula | C15H15N3O2 |
| Certificate of Analysis | Yes |
| pH Color Changes | Red: pH <4.4 Yellow: pH >6.2 Orange:pH 4.4–6.2 |
| Solvent or Matrix | Sodium hydroxide, alcohol and water |
| Color Index No. | 13020 |
| Synonym | C.I. Acid Red 2, 2-(4-Dimethylamino-phenylazo)benzoic acid |
| Show More |
Safety and Handling
Product Identifier
- METHYL RED SOLUTION
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: ethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
