missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red Solution, Solstice

Product Code. 15653860
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15653860 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15653860 Supplier Honeywell-Fluka Supplier No. 32941100ML
  • ONLINE EXCLUSIVE PRICE
    76.02 €
    64.00 € / 100mL
    Save 12.02 €
    15% Off
 Restricted Item
Estimated Shipment: 26-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acid-base indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 493-52-7
Molecular Formula C15H15N3O2
MDL Number MFCD00002425
UN Number 1170
Beilstein 1843037
Quantity 100 mL
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
Molecular Weight (g/mol) 269.304
PubChem CID 10303
Formula Weight 269.3g/mol
Chemical Name or Material Methyl Red Solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl Red solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.