missing translation for 'onlineSavingsMsg'
Learn More

Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%

Product Code. p-7041432
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437016 1 g 1g
15447016 5 g 5g
15457016 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15437016 Supplier Thermo Scientific Alfa Aesar Supplier No. H62091.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 108963-96-8 | C11H17NO5 | 243.259 g/mol

Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate in medicine, pharmacy, cosmetics, and industrial, artificial flavoring and fragrance agents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate in medicine, pharmacy, cosmetics, and industrial, artificial flavoring and fragrance agents

Solubility
Soluble in dichloromethane.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108963-96-8
Molecular Formula C11H17NO5
Molecular Weight (g/mol) 243.259
MDL Number MFCD06809720
InChI Key FNTAOUUEQHKLIU-ZETCQYMHSA-N
Synonym boc-l-pyroglutamic acid methyl ester, boc-pyr-ome, s-1-tert-butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate, methyl l-n-boc-pyroglutamate, 1-tert-butyl 2-methyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, methyl boc-l-pyroglutamate, s-5-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 1-tert-butyl 2-methyl s-5-oxopyrrolidine-1,2-dicarboxylate, boc-pglu-ome, pubchem12135
PubChem CID 10868485
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S)-5-oxopyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1C(CCC1=O)C(=O)OC

Specifications

Melting Point 68°C to 72°C
Quantity 1 g
UN Number UN3077
Solubility Information Soluble in dichloromethane.
Optical Rotation −32° (c=0.5 in Chloroform)
Formula Weight 243.26
Percent Purity 98%
Chemical Name or Material Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.