missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Salicylate-13C6, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl Salicylate-13C6 |
| Molecular Formula | C213C6H8O3 |
| Quantity | 25 mg |
| Synonym | 2-(Methoxycarbonyl)phenol-13C6, 2-Carbomethoxyphenol-13C6, 2-Hydroxybenzoic acid methyl ester-13C6, Analgit-13C6, Anthrapole ND-13C6, Ben Gay-13C6, Exagien-13C6, Flucarmit-13C6, Holly oil-13C6, Methyl 2-hydroxybenzoate-13C6, Methyl ester 2-hydroxy benzoic acid-13C6, Methyl ester of 2-hydroxy benzoic acid-13C6, Methyl o-hydroxybenzoate-13C6, NSC 8204-13C6, PredaLure-13C6, Wintergreen oil-13C6, o-Hydroxybenzoic acid methyl ester-13C6 |
| InChI Formula | InChI=1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3/i2+1,3+1,4+1,5+1,6+1,7+1 |
| SMILES | [H][13C]1=[13C]([13C](O)=[13C]([13C]([H])=[13C]1[H])[H])C(OC)=O |
| IUPAC Name | methyl 2-hydroxybenzoate-1,2,3,4,5,6-13C6 |
| Molecular Weight (g/mol) | 158.1 |
| Formula Weight | 158.07 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?