missing translation for 'onlineSavingsMsg'
Learn More

Methyl Salicylate, TRC

Product Code. 30479771
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479771 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30479771 Supplier Toronto Research Chemicals Supplier No. TRCM331025100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Salicylic acid methyl ester
CAS 119-36-8
Purity Grade Notes GC
Molecular Formula C8 H8 O3
Quantity 100 g
Synonym Methyl salicylate, Methyl 2-hydroxybenzoate, 2-Hydroxybenzoic acid methyl ester, Salicylic acid, methyl ester (6CI,8CI), 2-(Methoxycarbonyl)phenol, 2-Carbomethoxyphenol, 2-Hydroxybenzoic acid methyl ester, Analgit, Anthrapole ND, Ben Gay, Exagien, Flucarmit, Methyl 2-hydroxybenzoate, Methyl ester 2-hydroxy benzoic acid, Methyl ester of 2-hydroxy benzoic acid, Methyl o-hydroxybenzoate, Methyl salicylate, NSC 8204, PredaLure, Wintergreen oil, o-Hydroxybenzoic acid methyl ester
InChI Formula InChI=1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3
SMILES COC(=O)c1ccccc1O
IUPAC Name methyl 2-hydroxybenzoate
Molecular Weight (g/mol) 152.15
Formula Weight 152.0473
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.