missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfone, 98%

Product Code. 10306820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
2.5kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10306820 5 g Glass Bottle 5g
10655942 500 g Plastic bottle 500g
11904561 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10306820 Supplier Thermo Scientific Acros Supplier No. 127780050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 67-71-0 | C2H6O2S | 94.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-71-0
Molecular Formula C2H6O2S
Molecular Weight (g/mol) 94.13
MDL Number MFCD00007566
InChI Key HHVIBTZHLRERCL-UHFFFAOYSA-N
Synonym dimethyl sulfone, methyl sulfone, sulfonylbismethane, dimethylsulfone, dimethyl sulphone, methane, sulfonylbis, sulphonylbismethane, methylsulfone, methanesulfonylmethane, sulfonyldimethane
PubChem CID 6213
ChEBI CHEBI:9349
IUPAC Name methylsulfonylmethane
SMILES CS(C)(=O)=O

Specifications

Melting Point 107.0°C to 111.0°C
Color White
Boiling Point 238.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula (CH3)2SO2
Quantity 5 g
Beilstein 01,289
Fieser 01,296; 02,157
Merck Index 15,3283
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: freely soluble in methanol,ethanol and acetone,sparingly soluble in ether
Formula Weight 94.13
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl sulfone
Show More Show Less
Product Identifier
  • Methyl sulfone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.