missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide, 99.8+%, for peptide synthesis, Thermo Scientific Chemicals

Produktkod. 10269743 Handla allt Thermo Scientific Chemicals Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Förpackningsstorlek:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging unitSize
10269743 500 mL Glass Bottle 500mL
10633722 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Regleringsinformation: En licens krävs för att köpa denna produkt; KUNGALIG FÖRESKRIFT AV DEN 12 APRIL 1974.
Produktkod. 10269743 Leverantör Thermo Scientific Acros Leverantörsnummer 354845000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy
Regleringsinformation: En licens krävs för att köpa denna produkt; KUNGALIG FÖRESKRIFT AV DEN 12 APRIL 1974.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifikationer

Melting Point 18.4°C
Color Colorless
Density 1.1000g/mL
Quantity 500 mL
Flash Point 87°C
Packaging Glass Bottle
Physical Form Liquid at 20°C
Refractive Index 1.4770 to 1.4800
Residue after Evaporation 0.001% max.
Assay Percent Range 99.8+%
Specific Gravity 1.1
Linear Formula (CH3)2SO
Viscosity 1.987 mPa.s (25°C)
Water 0.005% max. (K.F.)
Beilstein 01, IV, 1277
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Merck Index 15, 3285
Boiling Point 189.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
Formula Weight 78.13
Percent Purity ≥99.8%
Chemical Name or Material Methyl sulfoxide, For peptide synthesis
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.