missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D

Product Code. 10497000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10497000 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10497000 Supplier Thermo Scientific Acros Supplier No. 169020250
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 1.1900g/mL
Deuterium (D) 99.95% min.
Flash Point 88°C
Specific Gravity 1.19
Viscosity 2.4 mPa.s (20°C)
Water 0.02% max. (Coulometric)
Quantity 25 mL
CAS 2206-27-1
Melting Point 20.2°C
Boiling Point 190.0°C
Molecular Formula C2H6OS
Linear Formula (CD3)2SO
MDL Number MFCD00002090
Beilstein 01, IV, 1279
Fieser 13,124; 16,177
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
Molecular Weight (g/mol) 84.17
PubChem CID 75151
Formula Weight 84.18
Grade NMR
Chemical Name or Material Methyl sulfoxide-d6
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.