missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide-d6, for NMR, contains 0.03% TMS, 99.8 atom% D

Product Code. 10190512 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
50 mL
Packaging:
Glass Bottle
Unit Size:
10mL
25mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10190512 10 mL Glass Bottle 10mL
16338196 25 mL Glass Bottle 25mL
10059041 50 mL Glass Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10190512 Supplier Thermo Scientific Acros Supplier No. 360000100
  • ONLINE EXCLUSIVE PRICE
    66.75 €
    64.80 € / 10mL
    Save 1.95 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-27-1
Molecular Formula C2H6OS
Molecular Weight (g/mol) 84.17
MDL Number MFCD00002090
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
PubChem CID 75151
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Density 1.1900g/mL
Deuterium (D) 99.75% min.
Flash Point 88°C
Packaging Glass Bottle
Specific Gravity 1.19
Viscosity 2.4 mPa.s (20°C)
Water 0.03% max. (Coulometric)
Quantity 10 mL
Melting Point 20.2°C
Boiling Point 190.0°C
Beilstein 01, IV, 1279
Fieser 13,124; 16,177
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
Formula Weight 84.18
Grade NMR
Chemical Name or Material Methyl sulfoxide-d6, 99.8 atom%D
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.