missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D

Product Code. 16255880 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
7.5 mL
Unit Size:
7.50mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 16255880

Brand: Thermo Scientific Acros 464010075

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

C2D6OS | 84.18 g/

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.1900 g/mL
Flash Point 88°C
Specific Gravity 1.1900
Viscosity 2.4 mPa.s (20°C)
Quantity 7.5 mL
Melting Point 20.2°C
Boiling Point 190.0°C
CAS 2206-27-1
Molecular Formula C2D6OS
Solubility Information Solubility aq. soln.: soluble.
Molecular Weight (g/mol) 84.18
Grade NMR
Chemical Name or Material Methyl sulfoxide-d6
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.