missing translation for 'onlineSavingsMsg'
Learn More

Methyl tert-Butyl Ether (MTBE), Pesticide Grade For Laboratory Use and Pesticide Analysis, Chem Lab

Product Code. 15979356
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15979356 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15979356 Supplier Chem-Lab Supplier No. CL00.3911.1000
  • ONLINE EXCLUSIVE PRICE
    101.00 €
    53.60 € / 1 L
    Save 47.40 €
    46% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1634-04-4
Melting Point -108.6°C
Color Colorless
Density 0.74 g/mL
Boiling Point 55.3°C
Flash Point -28 °C
Assay Percent Range > 99.9%
Packaging Glass bottles
Molecular Formula C5H12O
Quantity 1 L
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
SMILES COC(C)(C)C
IUPAC Name 2-methoxy-2-methylpropane
Molecular Weight (g/mol) 88.15
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl tert-butyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
Supplemental information
  • MIXTURE LIST-Contain: Methyl tert-butyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.