missing translation for 'onlineSavingsMsg'
Learn More

Methyl Tetradecanoate-d27, TRC

Product Code. 30478563
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478563 10 mg 10mg
30465298 5 mg 5mg
30472542 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30478563 Supplier Toronto Research Chemicals Supplier No. TRCM22760110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434349-93-6
Molecular Formula C15 D27 H3 O2
Molecular Weight (g/mol) 269.5639
Synonym Tetradecanoic-d27 acid, methyl ester (9CI), Tetradecanoic Acid Methyl Ester-D27 (tetradecanoic-D27), Tetradecanoic acid-d27 methyl ester, Myristic acid-d27 methyl ester, Methyl myristate-d27, Methyl tetradecanoate-d27
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosadeuteriotetradecanoate
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)OC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H30O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17-2/h3-14H2,1-2H3/i1D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2
Formula Weight 269.3941
Chemical Name or Material Methyl Tetradecanoate-d27
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.