missing translation for 'onlineSavingsMsg'
Learn More

Methyl thiazole-2-carboxylate, 95%

Product Code. 15480219
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480219 1 g 1g
15400229 250 mg 250mg
15490219 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15480219 Supplier Thermo Scientific Alfa Aesar Supplier No. H63496.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 55842-56-3 | C5H5NO2S | 143.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55842-56-3
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.16
MDL Number MFCD18075580
InChI Key YRMZUOPMHMPNPG-UHFFFAOYSA-N
PubChem CID 7567660
IUPAC Name methyl 1,3-thiazole-2-carboxylate
SMILES COC(=O)C1=NC=CS1

Specifications

Quantity 1 g
Synonym methyl thiazole-2-carboxylate, methyl 2-thiazolecarboxylate, 2-thiazolecarboxylic acid methyl ester, methyl2-thiazolecarboxylate
Formula Weight 143.16
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl thiazole-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.