missing translation for 'onlineSavingsMsg'
Learn More

Methyl thiocyanate, 99%

Product Code. 11387499
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
Product Code. Quantity unitSize
11387499 100 mL 100mL
11367499 5 mL 5mL
11377499 25 mL 25mL
3 options
This item is not returnable. View return policy

Product Code. 11387499

Brand: Thermo Scientific Alfa Aesar H27879.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 556-64-9 | C2H3NS | 73.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-64-9
Molecular Formula C2H3NS
Molecular Weight (g/mol) 73.113
MDL Number MFCD00001830
InChI Key VYHVQEYOFIYNJP-UHFFFAOYSA-N
Synonym methyl sulfocyanate, methylrhodanid, thiocyanic acid, methyl ester, methyl rhodanate, methylthiokyanat, methane, thiocyanato, thiocyanatomethane, methylthiocyanate, thiocyanic acid methyl ester, ch3scn
PubChem CID 11168
ChEBI CHEBI:61112
IUPAC Name methyl thiocyanate
SMILES CSC#N

Specifications

Melting Point -5°C
Density 1.068
Boiling Point 131°C
Flash Point 38°C (100°F)
Odor Pungent
Refractive Index 1.468
Quantity 100 mL
UN Number UN2929
Sensitivity Moisture sensitive
Formula Weight 73.11
Percent Purity 99%
Chemical Name or Material Methyl thiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.