missing translation for 'onlineSavingsMsg'
Learn More

Methyl thioglycolate, 98%

Product Code. 10413965 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10413965

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2365-48-2 | C3H6O2S | 106.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2365-48-2
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00004873
InChI Key MKIJJIMOAABWGF-UHFFFAOYSA-N
Synonym methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el
PubChem CID 16907
IUPAC Name methyl 2-sulfanylacetate
SMILES COC(=O)CS

Specifications

Melting Point -24°C
Color Colorless
Density 1.1870g/mL
Boiling Point 156°C to 159°C (10.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Iodimetry)
Packaging Glass bottle
Linear Formula HSCH2CO2CH3
Refractive Index 1.4640 to 1.467
Quantity 100 g
Specific Gravity 1.187
Solubility Information Solubility in water: 40g/l water (20°C)
Formula Weight 106.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl Thioglycolate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl thioglycolate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.