missing translation for 'onlineSavingsMsg'
Learn More

Methyl thiophene-2-carboxylate, 97%

Product Code. 11410563
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410563 5 g 5g
11420563 25 g 25g
11430563 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410563 Supplier Thermo Scientific Alfa Aesar Supplier No. A12677.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5380-42-7 | C6H6O2S | 142.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5380-42-7
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.17
MDL Number MFCD00016895
InChI Key PGBFYLVIMDQYMS-UHFFFAOYSA-N
Synonym methyl 2-thiophenecarboxylate, methyl thenoate, methylthiophene-2-carboxylate, 2-thiophenecarboxylic acid methyl ester, 2-carbomethoxy thiophene, 2-thiophenecarboxylic acid, methyl ester, thiophenate methyl, 2-methoxycarbonyl thiophene, methyl 2-thiophene carboxylate, thiophenate-methyl
PubChem CID 79340
IUPAC Name methyl thiophene-2-carboxylate
SMILES COC(=O)C1=CC=CS1

Specifications

Density 1.23
Boiling Point 95°C to 97°C (15 mmHg)
Refractive Index 1.542
Quantity 5 g
Beilstein 111111
Formula Weight 142.18
Percent Purity 97%
Chemical Name or Material Methyl thiophene-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.