missing translation for 'onlineSavingsMsg'
Learn More

Methyl tiglate, 98%

Product Code. 11423984
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423984 250 g 250g
11403984 10 g 10g
11413984 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423984 Supplier Thermo Scientific Alfa Aesar Supplier No. A11964.30
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6622-76-0 | C6H10O2 | 114.14 g/mol

It is employed as a perfuming agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a perfuming agent in cosmetics industry.

Solubility
Soluble in alcohol, water 3813 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6622-76-0
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00016654
InChI Key YYJWBYNQJLBIGS-PLNGDYQASA-N
Synonym methyl tiglate, tiglic acid methyl ester, methyl 2-methylbut-2-enoate, methyl 2-methyl-2-butenoate, methyl e-2-methylcrotonate, methyl 2-methylcrotonate, methyl alpha-methylcrotonate, methyl trans-2-methylcrotonate, 2-butenoic acid, 2-methyl-, methyl ester, e, crotonic acid, 2-methyl-, methyl ester, e
PubChem CID 5323652
IUPAC Name methyl (E)-2-methylbut-2-enoate
SMILES COC(=O)C(\C)=C/C

Specifications

Density 0.95
Boiling Point 138°C to 139°C
Flash Point 39°C (102°F)
Refractive Index 1.437
Quantity 250 g
UN Number UN3272
Beilstein 1720455
Merck Index 14,9433
Solubility Information Soluble in alcohol,water 3813mg/L @ 25°C (est).
Formula Weight 114.15
Percent Purity 98%
Chemical Name or Material Methyl tiglate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.