missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-4-aminocyclohexanecarboxylate hydrochloride, 97%

Product Code. 15417576
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417576 1 g 1g
15437576 250 mg 250mg
15427576 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15417576 Supplier Thermo Scientific Alfa Aesar Supplier No. H62562.03
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 61367-07-5 | C8H16ClNO2 | 193.671 g/mol

Methyl trans-4-aminocyclohexanecarboxylate hydrochloride is generally used as a intermediate in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride is generally used as a intermediate in pharmaceutical and chemical research.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61367-07-5
Molecular Formula C8H16ClNO2
Molecular Weight (g/mol) 193.671
MDL Number MFCD08274538
InChI Key NHAYDXCUCXRAMF-UHFFFAOYSA-N
Synonym methyl trans-4-aminocyclohexanecarboxylate hydrochloride, methyl 4-aminocyclohexanecarboxylate hydrochloride, methyl cis-4-aminocyclohexanecarboxylate hydrochloride, cis-methyl 4-aminocyclohexanecarboxylate hydrochloride, methyl 4-aminocyclohexane-1-carboxylate hydrochloride, trans-4-amino-cyclohexanecarboxylic acid methyl ester hydrochloride, cyclohexanecarboxylic acid, 4-amino-, methyl ester, hydrochloride, trans-4-amino-cyclohexylcarboxylic acid methyl ester hydrochloride, trans-methyl 4-aminocyclohexanecarboxylate hydrochloride, 4-amino-cyclohexanecarboxylic acid methyl ester hydrochloride
PubChem CID 18521575
IUPAC Name methyl 4-aminocyclohexane-1-carboxylate;hydrochloride
SMILES COC(=O)C1CCC(CC1)N.Cl

Specifications

Solubility Information Soluble in water.
Quantity 1 g
Formula Weight 193.67
Percent Purity 97%
Chemical Name or Material Methyl trans-4-aminocyclohexanecarboxylate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.