missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-4-Bromo-2-butenoate (>85%), TRC

Product Code. 30478155
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478155 5 g 5g
30465287 1 g 1g
30483510 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478155 Supplier Toronto Research Chemicals Supplier No. TRCM2940055G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6000-00-6
Molecular Formula C5H7BrO2
Molecular Weight (g/mol) 179.01
Synonym (E)-4-Bromo-2-butenoic Acid Methyl Ester, (E)-4-Bromo-crotonic Acid Methyl Ester, (2E)-4-Bromo-2-butenoic Acid Methyl Ester, (E)-4-Bromobut-2-enoic Acid Methyl Ester, (E)-Methyl 4-Bromobut-2-enoate, (E)-Methyl 4-Bromocrotonate, Methyl (E)-4-Bromo-2-butenoate, Methyl (E)-4-Bromocrotonate, Methyl trans-4-Bromocrotonate, trans-4-Bromobut-2-enoic Acid Methyl Ester
IUPAC Name methyl (E)-4-bromobut-2-enoate
SMILES O=C(OC)/C=C/CBr

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H7BrO2/c1-8-5(7)3-2-4-6/h2-3H,4H2,1H3/b3-2+
Formula Weight 177.96
Chemical Name or Material Methyl trans-4-Bromo-2-butenoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.