missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl trans-Cinnamate-d5 (phenyl-d5), CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Methyl trans-Cinnamate-D5 (phenyl-D5) |
| CAS | 61764-82-7 |
| Molecular Formula | C10 2H5 H5 O2 |
| Quantity | 0.1 g |
| Synonym | 2-Propenoic acid, 3-(phenyl-d5)-, methyl ester, (E)- (9 CI), Methyl trans-Cinnamate-D5 (phenyl-D5), (E)-3-(Phenyl-d5)-2-propenoic acid methyl ester, (E)-Cinnamic acid methyl ester d5 (phenyl-d5) |
| InChI Formula | InChI=1 S/C10H10O2/c1-12-10(11)8-7-9-5-3-2-4-6-9/h2-8 H,1H3/b8-7+/i2D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(\C=C\C(=O)OC)c([2 H])c1[2 H] |
| IUPAC Name | methyl (E)-3-(2,3,4,5,6-pentadeuteriophenyl)prop-2-enoate |
| Molecular Weight (g/mol) | 167.22 |
| Formula Weight | 167.0995 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction