missing translation for 'onlineSavingsMsg'
Learn More

Methyl trifluoromethanesulfonate, 96%, Thermo Scientific Chemicals

Product Code. 11346281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle in Tin Can
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11346281 100 g Glass bottle 100g
10435271 25 g Glass Bottle in Tin Can 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346281 Supplier Thermo Scientific Acros Supplier No. 415011000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-27-7
Molecular Formula C2H3F3O3S
Molecular Weight (g/mol) 164.10
MDL Number MFCD00000409
InChI Key OIRDBPQYVWXNSJ-UHFFFAOYSA-N
Synonym methyl triflate, methyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, methyl ester, triflate ester, methyl trifluoromethane sulfonate, trifluoromethanesulfonic acid methyl ester, unii-7b25z22epv, ccris 1158, methyltrifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, methyl ester
PubChem CID 9526
IUPAC Name methyl trifluoromethanesulfonate
SMILES COS(=O)(=O)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.4500g/mL
Boiling Point 95.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CF3SO3CH3
Refractive Index 1.3250 to 1.3270
Quantity 100 g
Fieser 06,406; 12,329; 13,327; 17,191
Merck Index 15,6204
Specific Gravity 1.45
Formula Weight 164.11
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methyl trifluoromethanesulfonate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.