missing translation for 'onlineSavingsMsg'
Learn More

Methyl valerate, 99%

Product Code. 10301351 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10301351 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10301351 Supplier Thermo Scientific Acros Supplier No. 415050250
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 624-24-8 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Acidity 1mg KOH/g max.
CAS 624-24-8
Color Colorless
Density 0.8900g/mL
Boiling Point 126°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H12O2
Linear Formula CH3(CH2)3COOCH3
Refractive Index 1.3965 to 1.3985
MDL Number MFCD00009478
Quantity 25 g
Beilstein 02, 301
Specific Gravity 0.89
Synonym methyl valerate, pentanoic acid, methyl ester, methyl n-valerate, methyl valerianate, valeric acid, methyl ester, valeric acid methyl ester, unii-zw21jjj9vn, pentanoic acid methyl ester, fema no. 2752, zw21jjj9vn
Solubility Information Solubility in water: insoluble
InChI Key HNBDRPTVWVGKBR-UHFFFAOYSA-N
SMILES CCCCC(=O)OC
IUPAC Name methyl pentanoate
Molecular Weight (g/mol) 116.16
PubChem CID 12206
Formula Weight 116.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl valerate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl valerate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.