missing translation for 'onlineSavingsMsg'
Learn More

Methyl Vanillate, TRC

Product Code. 30477218
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477218 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477218 Supplier Toronto Research Chemicals Supplier No. TRCM33849010G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Vanillate
CAS 3943-74-6
Molecular Formula C9H10O4
Quantity 10 g
Synonym 4-Hydroxy-3-methoxybenzoic Acid Methyl Ester, Methyl Ester Vanillic Acid, 4-Hydroxy-3-methoxy Methyl Benzoate, 4-Hydroxy-3-methoxybenzoic Acid Methyl Ester, Methyl 3-methoxy-4-hydroxybenzoate, Methyl 4-hydroxy-3-methoxybenzoate, Methyl vanillate, NSC 74385
InChI Formula InChI=1S/C9H10O4/c1-12-8-5-6(9(11)13-2)3-4-7(8)10/h3-5,10H,1-2H3
SMILES COC(=O)c1ccc(O)c(OC)c1
IUPAC Name methyl 4-hydroxy-3-methoxybenzoate
Molecular Weight (g/mol) 182.17
Formula Weight 182.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.