missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Viologen-d14 Dichloride, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Paraquat-D14 Dichloride |
| Molecular Formula | C12 D14 N2 . 2 Cl |
| Quantity | 0.05 g |
| Synonym | 1,1'-Di(methyl-d3)-4,4'-bipyridinium-2,2',3,3',5,5',6,6'-d8 Dichloride, Paraquat-D14 Dichloride, Methyl Viologen-d14 Dichloride |
| InChI Formula | InChI=1 S/C12H14N2.2 ClH/c1-13-7-3-11(4-8-13)12-5-9-14(2)10-6-12;;/h3-10 H,1-2H3;2*1 H/q+2;;/p-2/i1D3,2D3,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D;; |
| SMILES | [Cl-].[Cl-].[2 H]c1c([2 H])[n+](c([2 H])c([2 H])c1c2c([2 H])c([2 H])[n+](c([2 H])c2[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-[2,3,5,6-tetradeuterio-1-(trideuteriomethyl)pyridin-1-ium-4-yl]-1-(trideuteriomethyl)pyridin-1-ium;dichloride |
| Molecular Weight (g/mol) | 271.2452 |
| Formula Weight | 270.1413 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction