missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Yellow, indicator

Product Code. 11424647
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424647 500 g 500g
11404647 25 g 25g
11414647 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424647 Supplier Thermo Scientific Chemicals Supplier No. B21145.36
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 60-11-7 | C14H15N3 | 225.30 g/mol

Methyl Yellow used as a pH indicator. It is an acid-base indicator that changes color from below pH 2.9 red to above pH 4.0 yellow and vice versa. Typical use is in the titration of a weak base with a strong acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Yellow used as a pH indicator. It is an acid-base indicator that changes color from below pH 2.9 red to above pH 4.0 yellow and vice versa. Typical use is in the titration of a weak base with a strong acid.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Transition interval: pH 2.9 (red) to pH 4.0 (yellow)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-11-7
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.30
MDL Number MFCD00008308
InChI Key JCYPECIVGRXBMO-UHFFFAOYSA-N
Synonym methyl yellow, dimethyl yellow, 4-dimethylamino azobenzene, 4-dimethylaminoazobenzene, solvent yellow 2, sudan yellow, fat yellow, p-dimethylaminoazobenzene, stear yellow jb, sudan yellow gg
PubChem CID 6053
ChEBI CHEBI:17903
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 110°C to 112°C (decomposition)
Odor Odorless
Quantity 500 g
UN Number UN2811
Beilstein 746016
Merck Index 14,3229
Solubility Information Insoluble in water; Soluble in alcohol.
IUPAC Name N,N-dimethyl-4-(2-phenyldiazen-1-yl)aniline
Formula Weight 225.3
Chemical Name or Material Methyl Yellow, indicator

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.