missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Yellow, pure, indicator grade

Product Code. 10193720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10193720 100 g Glass bottle 100g
10219183 25 g Glass bottle 25g
10170030 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10193720 Supplier Thermo Scientific Chemicals Supplier No. 151401000
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60-11-7 | C14H15N3 | 225.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-11-7
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.30
MDL Number MFCD00008308
InChI Key JCYPECIVGRXBMO-UHFFFAOYSA-N
Synonym 4-Dimethylaminoazobenzene, N, N-Dimethyl-4-phenylazoaniline, Solvent Yellow 2
PubChem CID 6053
ChEBI CHEBI:17903
IUPAC Name N,N-dimethyl-4-phenyldiazenylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 111.0°C to 117.0°C
Infrared Spectrum Authentic
Loss on Drying 3% max.
Packaging Glass bottle
Linear Formula (CH3)2NC6H4N=NC6H5
Solubility Information Solubility in water: 13.6 mg/l. Other solubilities: soluble ca. 140g/L in acetone, soluble in benzene, chloroform, diethyl ether,, ethanol, petroleum ether, vegetable oils, alcohol,, ether, strong mineral acids and soils
Quantity 100 g
Physical Form Powder
Formula Weight 225.29
Grade Pure
Chemical Name or Material Methyl Yellow
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl Yellow, indicator grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.