missing translation for 'onlineSavingsMsg'
Learn More

Methylamine Hydrochloride, TRC

Product Code. 30476464
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30476464 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30476464

Brand: Toronto Research Chemicals TRCM25887525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methanamine Hydrochloride
CAS 593-51-1
Purity Grade Notes HPLC
Molecular Formula C H5 N . HCl
Quantity 25 g
Synonym Methanamine, hydrochloride (1:1), Methanamine, hydrochloride (9CI), Methylamine, hydrochloride (8CI), Methanaminium chloride, Methylamine hydrochloride, Methylamine monohydrochloride, Methylammonium chloride, Monomethylammonium chloride
InChI Formula InChI=1S/CH5N.ClH/c1-2;/h2H2,1H3;1H
SMILES Cl.CN
IUPAC Name methanamine;hydrochloride
Molecular Weight (g/mol) 67.518
Formula Weight 67.0189
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.