missing translation for 'onlineSavingsMsg'
Learn More

Methylamine Hydroiodide, TRC

Product Code. 30477800
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477800 1 g 1g
30476322 10 g 10g
30465799 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477800 Supplier Toronto Research Chemicals Supplier No. TRCM2219451G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14965-49-2
Molecular Formula CH5NHI
Molecular Weight (g/mol) 158.97
Synonym Methanamine, hydriodide (9CI), Methylamine, hydriodide (8CI), Methylamine hydroiodide, Methylammonium iodide, Methanamine hydroiodide
IUPAC Name methanamine;hydroiodide
SMILES I.CN

Specifications

Quantity 1 g
InChI Formula InChI=1S/CH5N.HI/c1-2;/h2H2,1H3;1H
Formula Weight 158.9545
Chemical Name or Material Methylamine Hydroiodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.