missing translation for 'onlineSavingsMsg'
Learn More

Methylaminoacetonitrile hydrochloride, 98%

Product Code. 15441388
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441388 25 g 25g
15431388 5 g 5g
15451388 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441388 Supplier Thermo Scientific Alfa Aesar Supplier No. H55215.14
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25808-30-4 | C3H7ClN2 | 106.553 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25808-30-4
Molecular Formula C3H7ClN2
Molecular Weight (g/mol) 106.553
MDL Number MFCD00012587
InChI Key DFKBQHBEROHUNF-UHFFFAOYSA-N
Synonym methylaminoacetonitrile hydrochloride, 2-methylamino acetonitrile hydrochloride, acetonitrile, methylamino-, monohydrochloride, sarcosinenitrile hydrochloride, n-methylaminoacetonitrile hydrochloride, acetonitrile, 2-methylamino-, hydrochloride 1:1, methylamino acetonitrilhydrochlorid, methylaminoacetonitrile, hydrochloride, methylaminoacetonitrilehydrochloride, cyanomethyl methyl ammonium chloride
PubChem CID 117641
IUPAC Name 2-(methylamino)acetonitrile;hydrochloride
SMILES CNCC#N.Cl

Specifications

Melting Point 103°C to 106°C
Linear Formula CH3NHCH2CN·HCl
Quantity 25 g
Beilstein 3908975
Formula Weight 106.55
Percent Purity 99%
Chemical Name or Material Methylaminoacetonitrile hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.