missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Methylation Smittel TMSH

Product Code. 10389991 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
Unit Size:
Each
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10389991 1 mL Pack of 20
10123013 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10389991 Supplier Macherey-Nagel™ Supplier No. 701520.201
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Item Description Methylation Smittel TMSH
Quantity 1 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • TMSH,0,2 M, 20x1 mL
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P311-Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: trimethylsulfonium hydroxide (TMSH), methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.