missing translation for 'onlineSavingsMsg'
Learn More

Methylcellulose, TRC

Product Code. 30010235
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010235 10 g 10g
30005359 50 g 50g
30008595 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010235 Supplier Toronto Research Chemicals Supplier No. TRCM29494510G
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.5
Synonym Adulsin, Avicel SG, Bagolax, Benecel 143HR, Benecel M 0, Benecel M 011, Benecel M 02, Benecel M 021, Benecel M 043, Benecel MC 4000PS, Benecel MO 42, Bufapto Methalose, Bulkaloid, Celacol M;, Methocel 4C, Methocel 64625, Methocel A, Methocel A 100, Methocel A 125LV, Methocel A 15, Methocel A 15C, Methocel A 15C Premium, Methocel A 15FG, Methocel A 15LV, Methocel A 15LV Premium, Methocel A 40M, Methocel A 4C, Methocel A 4C Premium, Methocel A 4C-FG, Methocel A 4M, Methocel A 4M Premium, Methocel A 50, Methocel AC Premium, Methocel CHG, Methocel Fluka 64625, Methocel HPM 500DS, Methocel HPM 50DS, Metolose SM 100, Metolose SM 10000, Metolose SM 15, Metolose SM 1500, Metolose SM 25, Metolose SM 4, Metolose SM 400, Metolose SM 4000, Metolose SM 65-15, Metolose SM 800, Metolose SM 8000, Metolose SR, Napolone, Nicel, O-Methylcellulose, Probe D, Rhomellose, SBQ 30000PE, SM 100, SM 10000, SM 100000, SM 15Tylose SAP, Tylose SL, Tylose SL 100, Tylose SL 400, Tylose SL 600, Tylose TWA, V 32, V 32 (cellulose derivative), Viscol, Viscontran L 52, Viscontran MC 400, Viscosol, Viskontran MC 10S, Walocel M, Walocel M 20678, Walsroder MC 20000S, XT 6000
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C20H38O11/c1-21-9-11-13(23-3)15(24-4)18(27-7)20(30-11)31-14-12(10-22-2)29-19(28-8)17(26-6)16(14)25-5/h11-20H,9-10H2,1-8H3
Chemical Name or Material Methylcellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.