missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexane, 99%, extra pure

Product Code. 10031180 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1 L
2.5 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10031180 2.5 L Glass Bottle 2.5 L
10091200 1 L Glass Bottle 1 L
10225621 5 L Plastic drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10031180 Supplier Thermo Scientific Acros Supplier No. 126580025
  • ONLINE EXCLUSIVE PRICE
    80.15 €
    77.80 € / 2.5 L
    Save 2.35 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-87-2 | C7H14 | 98.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-87-2
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00001497
InChI Key UAEPNZWRGJTJPN-UHFFFAOYSA-N
Synonym cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
PubChem CID 7962
IUPAC Name methylcyclohexane
SMILES CC1CCCCC1

Specifications

Melting Point -126.3°C
Density 0.7700g/mL
Boiling Point 101.0°C
Flash Point −3°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula C6H11CH3
Refractive Index 1.4210 to 1.4230
Quantity 2.5 L
Beilstein 05, 29
Merck Index 15, 6118
Specific Gravity 0.77
Solubility Information Solubility in water: 0.1g/L (20°C). Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,ether,ethanol
Formula Weight 98.19
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Methylcyclohexane, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylcyclohexane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: Methylcyclohexane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.