missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexane, 99%, Spectrophotometric Grade, Solstice

Product Code. 15612800
Change view
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass bottle
Unit Size:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15612800 1 L Glass bottle 1 L
15622800 2 L Glass bottle 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15612800 Supplier Fluka Supplier No. 2596911L
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

Spectrophotometric Grade, 99%

  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-87-2
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00001497
InChI Key UAEPNZWRGJTJPN-UHFFFAOYSA-N
Synonym cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
PubChem CID 7962
IUPAC Name methylcyclohexane
SMILES CC1CCCCC1

Specifications

Melting Point -126°C
Density 0.77g/mL at 25°C
Boiling Point 100°C to 102°C
Flash Point −4°C (24.8°F)
Packaging Glass bottle
Linear Formula C6H11CH3
Refractive Index 1.422
Quantity 1 L
UN Number UN2296
Beilstein 505972
Formula Weight 98.19g/mol
Percent Purity 99%
Grade Sprectrophotometric
Chemical Name or Material Methylcyclohexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.