missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclopentane, 95%

Product Code. 10736443 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
Unit Size:
2.5 L
25 L
500mL
This item is not returnable. View return policy

Product Code. 10736443

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-37-7 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-37-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00001382
InChI Key GDOPTJXRTPNYNR-UHFFFAOYSA-N
Synonym cyclopentane, methyl, methyl cyclopentane, methylpentamethylene, unii-5g26cc1ask, methyl-cyclopentane, ccris 6058, hsdb 876, 5g26cc1ask, un2298 flammable liquid, 3-methylcyclopentyl
PubChem CID 7296
IUPAC Name methylcyclopentane
SMILES CC1CCCC1

Specifications

Melting Point -142.4°C
Color White
Density 0.7480g/mL
Boiling Point 71.0°C
Flash Point −4°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Metal Drum
Linear Formula C5H9CH3
Refractive Index 1.4077 to 1.4097
Quantity 25 L
Beilstein 05, 27
Specific Gravity 0.748
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,diethyl ether,ethanol,petroleum ether
Viscosity 0.505 mPa.s (20°C)
Formula Weight 84.16
Percent Purity 95%
Physical Form Adhering Crystals
Chemical Name or Material Methylcyclopentane, 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylcyclopentane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.