missing translation for 'onlineSavingsMsg'
Learn More

Methyldiglycol, Extra Pure, SLR, Fisher Chemical

Product Code. 10366380 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10366380 1 L Amber glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10366380 Supplier Fisher Chemical Supplier No. M/4720/15
  • ONLINE EXCLUSIVE PRICE
    579.00 €
    571.00 € / 1 L
    Save 8.00 €
    1% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 111-77-3
Melting Point -70°C
Color Colorless
Boiling Point 194°C
Packaging Amber glass bottle
Molecular Formula C5H12O3
MDL Number MFCD00002871,MFCD00084416
Quantity 1 L
Synonym 2-2-methoxyethoxy ethanol, diethylene glycol monomethyl ether, methyl carbitol, methyl digol, methoxydiglycol, ethanol, 2-2-methoxyethoxy, methyl dioxitol, diethylene glycol methyl ether, dowanol dm, poly-solv dm
InChI Key SBASXUCJHJRPEV-UHFFFAOYSA-N
SMILES COCCOCCO
IUPAC Name 2-(2-methoxyethoxy)ethanol
Molecular Weight (g/mol) 120.15
PubChem CID 8134
Vapor Density 4.1
ChEBI CHEBI:44836
Formula Weight 120.15g/mol
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Methyldiglycol
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.