missing translation for 'onlineSavingsMsg'
Learn More

Methyldiphenylsulfonium tetrafluoroborate, 95%

Product Code. 11380074
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380074 5 g 5g
11390074 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380074 Supplier Thermo Scientific Alfa Aesar Supplier No. L00512.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 10504-60-6 | C13H13BF4S | 288.11 g/mol

Methyldiphenylsulfonium tetrafluoroborate is a Methylene transfer agent. It is also used in alkyl diphenylsulfonium salts in place of trialkyl avoids sulfur-containing by-products arising from the Sommelet-Hauser rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyldiphenylsulfonium tetrafluoroborate is a Methylene transfer agent. It is also used in alkyl diphenylsulfonium salts in place of trialkyl avoids sulfur-containing by-products arising from the Sommelet-Hauser rearrangement.

Solubility
Soluble in water, alcohol.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10504-60-6
Molecular Formula C13H13BF4S
Molecular Weight (g/mol) 288.11
MDL Number MFCD00051861
InChI Key LHAMVDBAOJCBDW-UHFFFAOYSA-N
Synonym diphenyl methyl sulfonium tetrafluoroborate, methyldiphenylsulphonium tetrafluoroborate 1-, methyldiphenylsulfonium tetrafluoroborate, methyldiphenylsulfanium tetrafluoroborate, acmc-1bx8v, diphenyl methyl sulfoniumtetrafluoroborate, methyl diphenyl sulfanium tetrafluoroborate, diphenyl methyl sulphonium tetrafluoroborate, methyl diphenyl sulfonium tetrafluoroborate
PubChem CID 2758717
IUPAC Name methyl(diphenyl)sulfanium;tetrafluoroborate
SMILES F[B-](F)(F)F.C[S+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 63°C to 67°C
Linear Formula (C6H5)2SCH3BF4
Quantity 5 g
UN Number UN1759
Beilstein 4732019
Formula Weight 288.11
Percent Purity 95%
Chemical Name or Material Methyldiphenylsulfonium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.