missing translation for 'onlineSavingsMsg'
Learn More

Methyldopate, TRC

Product Code. 30467228
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30467228 2.5 g 2.5g
30472705 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30467228 Supplier Toronto Research Chemicals Supplier No. TRCM3038652.5G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2508-79-4
Molecular Formula C12 H17 N O4 . Cl H
Molecular Weight (g/mol) 275.73
Synonym L-Tyrosine, 3-hydroxy-α-methyl-, ethyl ester, hydrochloride (1:1), Alanine, 3-(3,4-dihydroxyphenyl)-2-methyl-, ethyl ester, hydrochloride, (-)- (8CI), L-Tyrosine, 3-hydroxy-α-methyl-, ethyl ester, hydrochloride (9CI), Aldomet ester hydrochloride, Methyldopa ethyl ester hydrochloride, Methyldopate hydrochloride, α-Methyl-L-dopa ethyl ester hydrochloride, α-Methyldopa ethyl ester hydrochloride
IUPAC Name ethyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate;hydrochloride
SMILES Cl.CCOC(=O)[C@@](C)(N)Cc1ccc(O)c(O)c1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C12H17NO4.ClH/c1-3-17-11(16)12(2,13)7-8-4-5-9(14)10(15)6-8;/h4-6,14-15H,3,7,13H2,1-2H3;1H/t12-;/m0./s1
Formula Weight 275.0924
Chemical Name or Material Methyldopate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.