missing translation for 'onlineSavingsMsg'
Learn More

Methyle1,2,3-benzothiadiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10292452
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10292452 250 mg Amber Glass Bottle 250mg
10324342 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10292452 Supplier Thermo Scientific Maybridge Supplier No. KM10174CB
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 23616-15-1
Molecular Formula C8H6N2O2S
Molecular Weight (g/mol) 194.208
MDL Number MFCD01566550
InChI Key JCRFJMHWEAYQAA-UHFFFAOYSA-N
Synonym methyl benzo d 1,2,3 thiadiazole-5-carboxylate, methyl1,2,3-benzothiadiazole-5-carboxylate, 5-carbomethoxy-1,2,3-benzothiadiazole, maybridge1_005151, 5-methoxycarbonyl-1,2,3-benzothiadiazole, methyl benzo 1,2,3 thiadiazole-5-carboxylate, methylbenzo d 1,2,3 thiadiazole-5-carboxylate, 4-methylbenzo d 1,2,3 thiadiazole-5-carboxylate, 1,2,3-benzothiadiazole-5-carboxylic acid methyl ester, 1,2,3-benzothiadiazole-5-carboxylicacid, methyl ester
PubChem CID 2736813
IUPAC Name methyl 1,2,3-benzothiadiazole-5-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)SN=N2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 194.21
Percent Purity 97%
Chemical Name or Material Methyle1, 2, 3-benzothiadiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.