missing translation for 'onlineSavingsMsg'
Learn More

Methyle1H-benzimidazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10317074
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10317074 10 g Amber Glass Bottle 10g
10213032 250 mg Amber Glass Bottle 250mg
10171312 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10317074 Supplier Thermo Scientific Maybridge Supplier No. MO00298EA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 26663-77-4
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD03407310
InChI Key WJHHIVYNOVTVGY-UHFFFAOYSA-N
Synonym methyl benzimidazole-5-carboxylate, methyl 1h-benzimidazole-5-carboxylate, methyl 1h-benzo d imidazole-5-carboxylate, methyl 1h-1,3-benzodiazole-5-carboxylate, 1h-benzimidazole-5-carboxylicacidmethylester, 1h-benzimidazole-5-carboxylic acid methyl ester, methyl 1h-benzo d imidazole-6-carboxylate, benzimidazole-5-carboxylic acid methyl ester, 1h-benzoimidazole-5-carboxylic acid methyl ester, 1h-benzimidazole-5-carboxylic acid, methyl ester
PubChem CID 2779733
IUPAC Name methyl 3H-benzimidazole-5-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)N=CN2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material Methyl 1H-benzimidazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.