missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-isothiocyanatobenzoate, 97%, Thermo Scientific™

Product Code. 10722037
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10722037 25 g 25g
10302315 5 g 5g
10743574 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10722037 Supplier Thermo Scientific Maybridge Supplier No. SB01695EB
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 16024-82-1
Molecular Formula C9H7NO2S
Molecular Weight (g/mol) 193.22
MDL Number MFCD00041062
InChI Key UNXVHBOJSCWVCD-UHFFFAOYSA-N
Synonym 2-isothiocyanato-benzoic acid methyl ester, 2-methoxycarbonylphenyl isothiocyanate, 2-methoxycarbonyl phenyl isothiocyanate, benzoic acid,2-isothiocyanato-, methyl ester, benzoic acid, 2-isothiocyanato-, methyl ester, acmc-209dkr, methyl2-isothiocyanatobenzoate, methyl 2-isothiocyanato benzoate, methyl 2-isothiocyanatobenzenecarboxylate, 2-isothiocyanatobenzoic acid methyl ester
PubChem CID 85243
IUPAC Name methyl 2-isothiocyanatobenzoate
SMILES COC(=O)C1=CC=CC=C1N=C=S

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula SCNC6H4CO2CH3
Quantity 25 g
Formula Weight 193.23
Percent Purity 97%
Chemical Name or Material Methyl 2-isothiocyanatobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.