missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-amino-4-(methylsulfonyl)thiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10364762
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10364762 10 g 10g
10161212 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10364762 Supplier Thermo Scientific Maybridge Supplier No. KM01956EA
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175201-73-7
Molecular Formula C7H9NO4S2
Molecular Weight (g/mol) 235.272
MDL Number MFCD00067932
InChI Key ZFMCGKFMQDQBLY-UHFFFAOYSA-N
PubChem CID 2778572
IUPAC Name methyl 3-amino-4-methylsulfonylthiophene-2-carboxylate
SMILES COC(=O)C1=C(C(=CS1)S(=O)(=O)C)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Synonym methyl 3-amino-4-methylsulfonyl thiophene-2-carboxylate, methyl 3-amino-4-methanesulfonylthiophene-2-carboxylate, methyl 3-amino-4-methylsulphonyl thiophene-2-carboxylate, 3-amino-4-methanesulfonyl-thiophene-2-carboxylic acid methyl ester, methyl 3-amino-4-methylsulfonyl thiophene, maybridge1_004594, timtec-bb sbb005527, buttpark 33\04-59, methyl3-amino-4-methylsulfonyl thiophene-2-carboxylate, methyl 3-amino-4-methylsulfonyl thiophene-2carboxylate
Formula Weight 235.28
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-4-(methylsulfonyl)thiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.