missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-amino-4-methylthiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10391865
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10391865 5 g 5g
10304782 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10391865 Supplier Thermo Scientific Maybridge Supplier No. SB01297DE
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 85006-31-1
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00051822
InChI Key YICRPERKKBDRSP-UHFFFAOYSA-N
Synonym methyl3-amino-4-methylthiophene-2-carboxylate, 3-amino-4-methylthiophene-2-carboxylic acid methyl ester, unii-3la24473as, 3-amino-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-4-methyl-2-thenoate, methyl 3-amino-4-methyl-2-thiophenecarboxylate, methyl-3-amino-4-methylthiophene-2-carboxylate, methyl 3-amino-4-methyl-thiophene-2-carboxylate, 3-amino-4-methyl-thiophene-2-carboxylic acid methyl ester
PubChem CID 123584
IUPAC Name methyl 3-amino-4-methylthiophene-2-carboxylate
SMILES COC(=O)C1=C(N)C(C)=CS1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 171.22
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-4-methylthiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.