missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-amino-5-phenylthiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10603023
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10603023 1 g Amber Glass Bottle 1g
10019411 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10603023 Supplier Thermo Scientific Maybridge Supplier No. GK00498DA
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 100063-22-7
Molecular Formula C12H11NO2S
Molecular Weight (g/mol) 233.285
MDL Number MFCD00068161
InChI Key QESSCNMSOLRYBO-UHFFFAOYSA-N
Synonym methyl 3-amino-5-phenyl-2-thiophenecarboxylate, 2-thiophenecarboxylic acid, 3-amino-5-phenyl-, methyl ester, methyl-3-amino-5-phenylthiophene-2-carboxylate, methyl 3-amino-5-4-phenyl thiophene-2-carboxylate, 3-amino-5-phenyl-thiophene-2-carboxylic acid methyl ester, maybridge1_003977, acmc-2097mh, methyl-3-amino-5-phenylthiophene-carboxylate, 3-amino-2-methoxycarbonyl-5-phenylthiophene, methyl 3-amino-5-phenyl-thiophene-2-carboxylate
PubChem CID 700562
IUPAC Name methyl 3-amino-5-phenylthiophene-2-carboxylate
SMILES COC(=O)C1=C(C=C(S1)C2=CC=CC=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 233.29
Percent Purity 97%
Chemical Name or Material Methyle3-amino-5-phenylthiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.