missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-amino-5-(tert-butyl)thiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10772863
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10772863 1 g 1g
10240135 5 g 5g
10743053 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10772863 Supplier Thermo Scientific Maybridge Supplier No. GK00671DA
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175137-03-8
Molecular Formula C10H15NO2S
Molecular Weight (g/mol) 213.295
MDL Number MFCD00068163
InChI Key WHJBGOQJMFAKHY-UHFFFAOYSA-N
Synonym methyl 3-amino-5-tert-butyl thiophene-2-carboxylate, 3-amino-5-tert-butyl-thiophene-2-carboxylic acid methyl ester, methyl 3-amino-5-4-tert-butyl thiophene-2-carboxylate, maybridge1_003935, methyl 3-amino-5-tert-butyl-2-thiophenecarboxylate, 3-amino-5-tert-butyl-2-thiophenecarboxylic acid methyl ester, 3-amino-5-tert-butylthiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylicacid, 3-amino-5-1,1-dimethylethyl-, methyl ester
PubChem CID 2777503
IUPAC Name methyl 3-amino-5-tert-butylthiophene-2-carboxylate
SMILES CC(C)(C)C1=CC(=C(S1)C(=O)OC)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 213.3
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-5-(tert-butyl)thiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.