missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-bromothiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10233232
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10233232 10 g Amber Glass Bottle 10g
10120652 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10233232 Supplier Thermo Scientific Maybridge Supplier No. SEW02691EA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 26137-08-6
Molecular Formula C6H5BrO2S
Molecular Weight (g/mol) 221.068
MDL Number MFCD00173839
InChI Key PEGSJNCGPSIJOX-UHFFFAOYSA-N
Synonym 3-bromothiophene-2-carboxylic acid methyl ester, 3-bromo-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-bromo-, methyl ester, 3-bromo-2-thiophenecarboxylic acid methyl ester, methyl 3-bromo-2-thiophenecarboxylate, maybridge3_006386, ksc494q2p, 2-methoxycarbonyl-3-bromothiophene, methyl-3-bromothiophene carboxylate, methyl 3-bromanylthiophene-2-carboxylate
PubChem CID 2740074
IUPAC Name methyl 3-bromothiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 221.07
Percent Purity 97%
Chemical Name or Material Methyle3-bromothiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.